Anti-TBX15 Rabbit Polyclonal Antibody

49-738
10079-268EA 811.46 CAD
10079-268
Anti-TBX15 Rabbit Polyclonal Antibody
T-box 15 antibody can be used in Western Blot, immunohistochemistry starting at 20 ?g/mL, and immunoprecipitation.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TBX15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR