Anti-TBX4 Rabbit Polyclonal Antibody

49-740
10079-272EA 736.85 CAD
10079-272
Anti-TBX4 Rabbit Polyclonal Antibody
T-box 4 antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 5 ?g/mL, and immunoprecipitation

PBS, 0.09% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TBX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR