Anti-TFRC Rabbit Polyclonal Antibody

49-742
10079-276EA 1011.8 CAD
10079-276
Anti-TFRC Rabbit Polyclonal Antibody
Antibodies
  • Transferrin Receptor antibody can be used in Western Blot, and immunohistochemistry starting at 1:100.

Protein G Column

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TFRC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR