Anti-THOC5 Rabbit Polyclonal Antibody

49-743
10079-278EA 811.46 CAD
10079-278
Anti-THOC5 Rabbit Polyclonal Antibody
THO Complex 5 antibody can be used in ELISA starting at 1:5000 - 1:25000, Western Blot starting at 1:500 - 1:2000, immunohistochemistry starting at 20 ?g/mL, immunocytochemistry starting at 1:100 - 1:500, and immunoprecipitation.

PBS, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: THOC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Frog, Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR