Anti-SAA1 Mouse Monoclonal Antibody

49-762
10079-306EA 811.46 CAD
10079-306
Anti-SAA1 Mouse Monoclonal Antibody
Recognizes natural and recombinant human SAA. Does not show any crossreactivity with other human cytokines or growth factors tested such as IL-1b, IL-8, MCAF, TGF-b and EGF.

SAA1 antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, pH 7.2, 40% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SAA1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR