Anti-CYCS Mouse Monoclonal Antibody [clone: [7H8.2C12]]

Supplier: Prosci

49-765
10074-510EA 955.07 CAD
10074-510
Anti-CYCS Mouse Monoclonal Antibody [clone: [7H8.2C12]]
Antibodies
In addition to its well-known role in electron transport, cytochrome c (CYT) has been identified as a mediator of apoptosis or programmed cell death. Cytochrome c translocates from mitochondria into the cytoplasm in the early phases of apoptosis and plays a role in initiation of the apoptotic proteolytic cascade by activating caspase-3. Cytochrome c has been shown to bind Apaf-1. Apaf-1 has also been shown to activate capase-3.

Somatic Cytochrome C antibody can be used in ELISA, immunohistochemistry starting at 5 ?g/mL, immunocytochemistry, immunofluorescence, and immunoprecipitation.

Protein G Column

PBS, 0.2% gelatin, 0.05% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CYCS
Clonality: Monoclonal
Clone: [7H8.2C12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Drosophila, Hamster, Horse, Human, Mouse, Pigeon, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR