Anti-COL18A1 Mouse Monoclonal Antibody

Supplier: Prosci

49-791
10074-546EA 1047.24 CAD
10074-546
Anti-COL18A1 Mouse Monoclonal Antibody
Antibodies
Suitable for use in Immunohistochemistry and Immunoblot. Other applications not tested. Recognizes endostatin, Mr 22kD, an unidentified protein is also weakly detected, Mr 54kD. Species Crossreactivity: Human and mouse. Predicted to crossreact with rat.

COL18A1 antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

Thiophilic Adsorption Chromatography

50 mM sodium phosphate, pH 8.0, 0.05% sodium azide, before the addition of glycerol to 30%

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: COL18A1
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR