Anti-CS Mouse Monoclonal Antibody

49-795
10074-550EA 906.92 CAD
10074-550
Anti-CS Mouse Monoclonal Antibody
In Western blot experiments suitable for non-reducing conditions only.

Citrate Synthase antibody can be used in immunohistochemistry starting at 5 ?g/mL.

Protein G Column

PBS, pH 7.6, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CS
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR