Anti-CK19 Mouse Monoclonal Antibody [clone: [C-11]]

49-798
10074-556EA 1011.8 CAD
10074-556
Anti-CK19 Mouse Monoclonal Antibody [clone: [C-11]]
Antibodies
Cytokeratins are a member of intermediate filaments subfamily represented in epithelial tissues.

Keratin antibody can be used in Western Blot starting at 1:200, and immunohistochemistry starting at 10 ?g/mL.

Protein A Column

PBS, 15 mM sodium azide, approx., pH 7.4.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: KRT19
Clonality: Monoclonal
Clone: [C-11]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mammal
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR