Anti-PHB2 Rabbit Polyclonal Antibody

Supplier: Prosci

49-844
10076-526EA 955.07 CAD
10076-526
Anti-PHB2 Rabbit Polyclonal Antibody
Antibodies
REA (repressor of estrogen receptor activity) has been identified as a coregulator that directly interacts with estrogen receptor (ER) and represses ER transcriptional activity. Suitable for use in Immunoblot, Immunoprecipitation and Immunocytochemistry. Other applications not tested. Recognizes REA, Mr 37kD. Species Crossreactivity: Mouse, human and rat.

Prohibitin 2 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

Protein A Column

Tris-glycine buffer, 15M NaCl, pH 7.4, 0.05% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: PHB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR