Anti-INS Mouse Monoclonal Antibody [clone: [8E2]] (HRP (Horseradish Peroxidase))

Supplier: Prosci

49-910
10076-570EA 955.07 CAD
10076-570
Anti-INS Mouse Monoclonal Antibody [clone: [8E2]] (HRP (Horseradish Peroxidase))
Antibodies
After removal of the precursor signal peptide, proinsulin is post-translationally cleaved into two chains (peptide A and peptide B) that are covalently linked via two disulfide bonds. Binding of this mature form of insulin to the insulin receptor (INSR) stimulates glucose uptake. A variety of mutant alleles with changes in the coding region have been identified.

Insulin antibody can be used in immunohistochemistry starting at 10 - 20 ?g/mL.

Protein A Column

PBS, pH 7.4, 0.05% Proclin 300

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: INS
Clonality: Monoclonal
Clone: [8E2]
Conjugation: HRP
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR