Anti-TF Rabbit Polyclonal Antibody

49-935
10079-320EA 620.52 CAD
10079-320
Anti-TF Rabbit Polyclonal Antibody
Transferrin (TF) is a glycoprotein with an approximate molecular weight of 76.5 kDa. TF is thought to have been created as a result of an ancient gene duplication event that led to generation of homologous C and N-terminal domains each of which binds 1 ion of ferric iron. The function of transferrin is to transport iron from the intestine, reticuloendothelial system, and liver parenchymal cells to all proliferating cells in the body. In addition to its function in iron transport, transferrin may also have a physiologic role as granulocyte/pollen-binding protein (GPBP) involved in the removal of certain organic matter/allergins from serum.

Serotransferrin antibody can be used in ELISA, and immunohistochemistry starting at 5 ?g/mL.

Ion Exchange Chromatography

PBS, pH 7.2, 0.01% sodium azide, 40% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: TF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR