Suitable for use in ELISA, Immunohistochemistry and Western Blot. Other applications not tested. Appears to recognize the human pre-synaptic protein, SNAP-25; an antigen of 26-27kD molecular weight. It will recognize SNAP-25 fusion protein from COS cells, but not with the fusion protein from bacterial systems. Research studies have used this antibody to study the distribution of synaptic changes in the hippocampus of patients with medically refractory temporal lobe epilepsy. (1., 3., 8.). Species Crossreactivity: Rat, hamster and porcine
SNAP25 antibody can be used in immunohistochemistry starting at 20 ?g/mL.
Protein A Column
PBS, pH 7.2, 0.09% sodium azide
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: Primary
Antigen: SNAP25
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Hamster, Human, Pig, Rat