Anti-OSTC Rabbit Polyclonal Antibody

Supplier: Prosci
49-989
10079-366EA 955.07 CAD
10079-366
Anti-OSTC Rabbit Polyclonal Antibody
Antibodies
OSTC antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: OSTC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chicken, Dog, Xenopus, Gorilla, Hamster, Human, Mouse, Pig, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR