Anti-ACTB Rabbit Polyclonal Antibody

Supplier: Prosci

50-110
10075-706EA 938.13 CAD
10075-706
Anti-ACTB Rabbit Polyclonal Antibody
Antibodies
Neat Serum

Type: Primary
Antigen: ACTB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR