Anti-SLC6A2 Mouse Polyclonal Antibody

50-127
10075-590EA 728.19 CAD
10075-590
Anti-SLC6A2 Mouse Polyclonal Antibody
Protein G purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Type:
Antigen: SLC6A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR