Anti-PROX1 Mouse Polyclonal Antibody [clone: 5G10]

Supplier: Prosci

50-140
10075-692EA 1054.94 CAD
10075-692
Anti-PROX1 Mouse Polyclonal Antibody [clone: 5G10]
Antibodies
Protein G purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Type: Primary
Antigen: PROX1
Clonality: Monoclonal
Clone: 5G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR