Anti-CHAT Rabbit Polyclonal Antibody

Supplier: Prosci
50-152
10075-528EA 937.89 CAD
10075-528
Anti-CHAT Rabbit Polyclonal Antibody
Antibodies
Neat serum

Type: Primary
Antigen: CHAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat, Non-Human Primate
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR