Anti-RPS6KA2 Rabbit Polyclonal Antibody

50-189
10075-440EA 587.93 CAD
10075-440
Anti-RPS6KA2 Rabbit Polyclonal Antibody
The p90 ribosomal S6 kinases (RSK)1–4 are downstream members of the extracellular signal-regulated kinase (ERK)/MAPK cascade. The loss of RSK2 activity in humans leads to Coffin–Lowry syndrome, which is characterized by mental retardation and growth deficits (Hanauer and Young, 2002). Recent work suggests that RSK2 exerts a tonic regulation on G-protein coupled signaling (Sheffler et al., 2006).

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RPS6KA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Bovine, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR