Anti-SYN1 Rabbit Polyclonal Antibody

Supplier: Prosci

50-191
10075-480EA 937.91 CAD
10075-480
Anti-SYN1 Rabbit Polyclonal Antibody
Antibodies
Synapsin I plays a key role in synaptic plasticity in the brain (Feng et al.,
2002; Nayak et al., 1996). This effect is due in large part to the ability of the synapsins to regulate
the availability of synaptic vesicles for release. In addition to its role in plasticity, the expression of
synapsin I is a precise indicator of synapse formation (Moore and Bernstein, 1989; Stone et al.,
1994). Thus, Synapsin I immunocytochemistry provides a valuable tool for the study of
synaptogenesis. The role of synapsin in synaptic plasticity and in synaptogensis is regulated by
phosphorylation (Jovanovic et al., 2001; Kao et al., 2002).

Neat Serum

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: SYN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR