Anti-PPP1R1B Rabbit Polyclonal Antibody

Supplier: Prosci

50-201
10075-704EA 1047.29 CAD
10075-704
Anti-PPP1R1B Rabbit Polyclonal Antibody
Antibodies
DARPP-32 is a dopamine (DA) and cAMP-regulated ~32k phosphoprotein that is associated with dopaminoceptive neurons (Fienberg et al., 1998). The protein inhibits Protein Phosphatase I when it is phosphorylated on Thr34. In contrast, when DARPP-32 is phosphorylated on Thr75 the protein acts as an inhibitor of PKA (Bibb et al., 1999). Phosphorylation of DARPP-32 is thought to play a critical role in the regulation of dopaminergic neurotransmission. In addition, the activity of DARPP-32 is also thought to play important roles
in the actions of alcohol, caffeine and Prozac® (Maldve et al., 2002; Lindskog et al., 2002; Svenningsson et al., 2002).

The antibody has been directly tested for reactivity in Western blots with rat tissue. It is anticipated that the antibody will also react with bovine, canine, chicken, human, mouse, rat, Xenopus and zebra fish based on the fact that these species have 100% homology with the amino acid sequence used as antigen.

Affinity Purified

100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: Ppp1r1b
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish, Bovine, Chcicken, Xenopus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR