Anti-NEFL Mouse Monoclonal Antibody [clone: DA2]

Supplier: Prosci

50-255
10075-412EA 1047.23 CAD
10075-412
Anti-NEFL Mouse Monoclonal Antibody [clone: DA2]
Antibodies
Neurofilaments are the 10nm or intermediate filament proteins specifically in neurons, and are composed predominantly of three major proteins called NF-
NF-M and NF-H. NF-L is the neurofilament light or low molecular weight polypeptide and runs on SDS-PAGE gels at about 68 kDa. Antibodies to NF-L are useful for identifying neuronal cells and their processes in tissue sections and in tissue culture. Mutations in the protein region of the human NF-L gene cause some forms of Charcot-Marie-Tooth disease.

Total IgG fraction

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: NEFL
Clonality: Monoclonal
Clone: DA2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR