Anti-GHR Mouse Monoclonal Antibody

51-005
10074-574EA 764.4 CAD
10074-574
Anti-GHR Mouse Monoclonal Antibody
GHR antibody can be used in Western Blot starting at 1 - 2 ?g/mL, immunohistochemistry starting at 5 ?g/mL, immunoprecipitation, and immunofluorescence.

PBS, pH 7.2, 0.1% sodium azide.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: GHR
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR