Anti-PLIN2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))

51-007
10074-578EA 753.5 CAD
10074-578
Anti-PLIN2 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate))
Adimmunoprecipitationophilin antibody can be used in immunohistochemistry starting at 5 ?g/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: PLIN2
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR