Anti-TNFRSF1A Rabbit Polyclonal Antibody

Supplier: Prosci

51-016
10074-592EA 1124.21 CAD
10074-592
Anti-TNFRSF1A Rabbit Polyclonal Antibody
Antibodies
TNFRSF1A antibody can be used in ELISA, Western Blot, immunohistochemistry starting at 10 ?g/mL, and immunofluorescence starting at 10 ?g/mL.

PBS, pH 7.2, 30% glycerol, 0.5% BSA, 0.01% Thimerosal.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: TNFRSF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Hamster, Human, Monkey, Mouse, Pig, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR