Anti-INS Rabbit Polyclonal Antibody

51-029
10079-374EA 736.85 CAD
10079-374
Anti-INS Rabbit Polyclonal Antibody
Insulin antibody can be used in immunohistochemistry starting at 10 - 20 ?g/mL.

10 mM HEPES, pH 7.5, 150 mM NaCl, 100 ug/ml BSA, 50% glycerol.

Store Insulin antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: INS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR