Anti-ITPR1 Rabbit Polyclonal Antibody

Supplier: Prosci

51-064
10076-608EA 1011.53 CAD
10076-608
Anti-ITPR1 Rabbit Polyclonal Antibody
Antibodies
ITPR1 antibody can be used in immunohistochemistry starting at 6.5 ?g/mL.

PBS, 0.01% sodium azide, 50% glycerol.

Store ITPR1 antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ITPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR