Anti-ALPP Rabbit Polyclonal Antibody

Supplier: Prosci

51-087
10076-648EA 955.07 CAD
10076-648
Anti-ALPP Rabbit Polyclonal Antibody
Antibodies
ALPP antibody can be used in ELISA, Western Blot, and immunohistochemistry starting at 5 ?g/mL.

20 mM Tris-HCl, pH 8.0, 20 mg/mL BSA, 0.05% sodium azide

Store ALPP antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ALPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR