Anti-LPA Sheep Polyclonal Antibody

51-103
10079-424EA 623.55 CAD
10079-424
Anti-LPA Sheep Polyclonal Antibody
Apolipoprotein A (apo A) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24,000 kDa. Apo A is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.

Limmunoprecipitationoprotein antibody can be used in ELISA starting at 1:10000, Western Blot starting at 1:500 - 1:2000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4, 0.05% sodium azide.

Store Lipoprotein antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: LPA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR