Anti-IAPP Rabbit Polyclonal Antibody

Supplier: Prosci

51-112
10079-440EA 1011.8 CAD
10079-440
Anti-IAPP Rabbit Polyclonal Antibody
Antibodies
Amylin is a pancreatic islet peptide with a role in the maintenance of glucose homoeostasis. It is predominantly found in the beta cells of the pancreas and to a lesser extent in the gastrointestinal tract and nervous system. This antibody may prove of value in studies to determine the level of staining for amylin in normal and diabetic pancreas and pancreatic carcinoma.

IAPP antibody can be used in Western Blot starting at 2 ?g/mL, immunohistochemistry starting at 20 ?g/mL, and flow cytometry.

10 mM PBS, pH 7.4, BSA, sodium azide.

Store IAPP antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: IAPP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR