Anti-IL18 Rabbit Polyclonal Antibody

Supplier: Prosci

51-130
10074-634EA 1011.8 CAD
10074-634
Anti-IL18 Rabbit Polyclonal Antibody
Antibodies
IL-18 antibody can be used in ELISA, Western Blot starting at 1:500 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2, 30% glycerol, 0.5% BSA, 0.01% Thimerosal.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: IL18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR