Anti-IL7R Rabbit Polyclonal Antibody

51-139
10074-652EA 623.55 CAD
10074-652
Anti-IL7R Rabbit Polyclonal Antibody
IL-7 Receptor antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.02 M potassium phosphate, 0.15 M NaCl, pH 7.2, 0.01% sodium azide.

Store IL-7 Receptor antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: IL7R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR