Anti-GPR88 Rabbit Polyclonal Antibody

Supplier: Prosci

51-247
10076-688EA 1011.8 CAD
10076-688
Anti-GPR88 Rabbit Polyclonal Antibody
Antibodies
GPR88 antibody can be used in immunohistochemistry starting at 20 ?g/mL.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: GPR88
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Human, Mouse, Rabbit, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR