Anti-LPCAT1 Rabbit Polyclonal Antibody

Supplier: Prosci
51-263
10074-660EA 955.07 CAD
10074-660
Anti-LPCAT1 Rabbit Polyclonal Antibody
Antibodies
LPCAT1 antibody can be used in Western Blot, immunohistochemistry starting at 10 ?g/mL, immunocytochemistry, immunoprecipitation, and flow cytometry.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: LPCAT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Chicken, Dog, Gorilla, Hamster, Human, Monkey, Mouse, Pig, Rat, Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR