Anti-TFAP2A Mouse Monoclonal Antibody

51-269
10074-672EA 620.52 CAD
10074-672
Anti-TFAP2A Mouse Monoclonal Antibody
AP2 Alpha antibody can be used in Western Blot, and immunohistochemistry starting at 1 - 2 ?g/mL.

PBS, pH 7.4, 0.09% sodium azide.

Store AP2 Alpha antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: TFAP2A
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR