Anti-ITGB2 Mouse Monoclonal Antibody [clone: [MEM-48]]

Supplier: Prosci

51-272
10074-678EA 1011.8 CAD
10074-678
Anti-ITGB2 Mouse Monoclonal Antibody [clone: [MEM-48]]
Antibodies
CD18 antibody can be used in Western Blot starting at 1 - 3 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

Phosphate buffered saline (PBS), approx. pH 7.4, 0.2 µm filter sterilized

CD18 antibody can be stored short term 4 °C. For long term storage aliquot and store at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ITGB2
Clonality: Monoclonal
Clone: [MEM-48]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR