Anti-ICAM3 Mouse Monoclonal Antibody

51-274
10074-682EA 736.85 CAD
10074-682
Anti-ICAM3 Mouse Monoclonal Antibody
ICAM3 antibody can be used in Western Blot starting at 0.5 - 2 ?g/mL, and immunohistochemistry starting at 10 ?g/mL.

Azide free phosphate buffered saline (PBS), approx. pH 7.4; 0.2 mm filter sterilized.

ICAM3 antibody should be stored at -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type:
Antigen: ICAM3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR