Anti-DDR1 Mouse Monoclonal Antibody [clone: [2G4E12]]

Supplier: Prosci

51-299
10074-692EA 1047.24 CAD
10074-692
Anti-DDR1 Mouse Monoclonal Antibody [clone: [2G4E12]]
Antibodies
DDR1 antibody can be used in ELISA starting at 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: DDR1
Clonality: Monoclonal
Clone: [2G4E12]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR