Anti-CANX Mouse Monoclonal Antibody [clone: [3H4A7]]

Supplier: Prosci

51-308
10076-718EA 955.07 CAD
10076-718
Anti-CANX Mouse Monoclonal Antibody [clone: [3H4A7]]
Antibodies
Calnexin antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CANX
Clonality: Monoclonal
Clone: [3H4A7]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR