Anti-CD38 Mouse Monoclonal Antibody [clone: [6E12D]]

Supplier: Prosci

51-310
10076-722EA 1011.8 CAD
10076-722
Anti-CD38 Mouse Monoclonal Antibody [clone: [6E12D]]
Antibodies
CD38 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: [6E12D]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR