Anti-MMP2 Mouse Monoclonal Antibody

51-332
10076-766EA 620.52 CAD
10076-766
Anti-MMP2 Mouse Monoclonal Antibody
MMP2 antibody can be used in ELISA, Western Blot starting at 0.5 - 2 ?g/mL, immunohistochemistry starting at 10 ?g/mL, and immunoprecipitation

Phosphate-buffered solution, pH 7.2, 0.09% sodium azide at 0.5 mg/ml.

Store MMP2 antibody at 4 °C. Store undiluted.

Type:
Antigen: MMP2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Gibbon, Dog, Gorilla, Horse, Human, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR