Anti-ITGB3 Mouse Monoclonal Antibody [clone: [VI-PL2]]

Supplier: Prosci

51-342
10076-784EA 1047.24 CAD
10076-784
Anti-ITGB3 Mouse Monoclonal Antibody [clone: [VI-PL2]]
Antibodies
ITGB3 antibody can be used in Western Blot starting at 1:2000, immunohistochemistry starting at 1:100, and immunoprecipitation.

PBS, pH 7.2, \<=0.09% sodium azide

Store ITGB3 antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: [VI-PL2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR