Anti-CCR7 Mouse Monoclonal Antibody

51-356
10076-794EA 656.81 CAD
10076-794
Anti-CCR7 Mouse Monoclonal Antibody
CCR7 antibody can be used in ELISA, Western Blot starting at 1 ?g/mL, and immunohistochemistry starting at 10 - 20 ?g/mL.

PBS, pH 7.2, 150 mM NaCl, 0.09% sodium azide

Store CR7 antibody at 4 °C. As with all antibodies avoid freeze/thaw cycles

Type:
Antigen: CCR7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR