Anti-RTN3 Mouse Monoclonal Antibody [clone: [10A8]]

Supplier: ProSci

51-381
10076-844EA 1011.8 CAD
10076-844
Anti-RTN3 Mouse Monoclonal Antibody [clone: [10A8]]
Antibodies
Reticulon 3 antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: RTN3
Clonality: Monoclonal
Clone: [10A8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR