Anti-FABP2 Mouse Monoclonal Antibody [clone: [9A9B7B3]]

Supplier: ProSci
51-388
10076-858EA 1011.8 CAD
10076-858
Anti-FABP2 Mouse Monoclonal Antibody [clone: [9A9B7B3]]
Antibodies
FABP2 antibody can be used in ELISA, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: FABP2
Clonality: Monoclonal
Clone: [9A9B7B3]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR