Anti-ERN1 Mouse Monoclonal Antibody [clone: [9F2]]

Supplier: ProSci

51-402
10076-884EA 955.07 CAD
10076-884
Anti-ERN1 Mouse Monoclonal Antibody [clone: [9F2]]
Antibodies
IRE1 antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

0.03% sodium azide

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type: Primary
Antigen: ERN1
Clonality: Monoclonal
Clone: [9F2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR