Anti-ITIH4 Mouse Monoclonal Antibody

Supplier: ProSci

51-416
10076-912EA 1011.53 CAD
10076-912
Anti-ITIH4 Mouse Monoclonal Antibody
Antibodies
Trypsin antibody can be used in ELISA starting at 1:000 - 1:1000, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.4

Store Trypsin antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.

Type: Primary
Antigen: ITIH4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR