Anti-ART3 Mouse Monoclonal Antibody [clone: [3A2]]

Supplier: ProSci

51-425
10076-928EA 1124.21 CAD
10076-928
Anti-ART3 Mouse Monoclonal Antibody [clone: [3A2]]
Antibodies
ART3 antibody can be used in Western Blot, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store ART3 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ART3
Clonality: Monoclonal
Clone: [3A2]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR