Anti-ATR Mouse Monoclonal Antibody [clone: [1E9]]

Supplier: ProSci
51-429
10076-936EA 1061.16 CAD
10076-936
Anti-ATR Mouse Monoclonal Antibody [clone: [1E9]]
Antibodies
ATR antibody can be used in Western Blot, and immunohistochemistry starting at 10 ?g/mL.

PBS, pH 7.2

Store ATR antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: ATR
Clonality: Monoclonal
Clone: [1E9]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR