Anti-RUNX2 Mouse Monoclonal Antibody [clone: [1D8]]

51-433
10074-720EA 906.92 CAD
10074-720
Anti-RUNX2 Mouse Monoclonal Antibody [clone: [1D8]]
RUNX2 antibody can be used in Western Blot starting at 1 - 2 ?g/mL, and immunohistochemistry starting at 5 ?g/mL.

PBS, pH 7.2

Store RUNX2 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.

Type: Primary
Antigen: RUNX2
Clonality: Monoclonal
Clone: [1D8]
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b,k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR